Cyclopropanecarboxamidine hydrochloride ≥98.0% (by total nitrogen basis)
Proveedor: TCI
|
Atención
|
C2344-1G
C2344-5G
TCIAC2344-1GEA
16
EUR
TCIAC2344-1G
TCIAC2344-5G
Cyclopropanecarboxamidine hydrochloride ≥98.0% (by total nitrogen basis)
Cyclopropanecarboxamidine hydrochloride
Formula:
C₄H₈N₂ |
Número MDL:
MFCD00053010 Núm. CAS: 57297-29-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 125.0 to 129.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...