6-Chloro-3-indolyl-β-D-galactopyranoside ≥98.0% (by HPLC) for biochemical research
Proveedor: TCI
Sinónimos:
Rose-Gal, Salmon-Gal
C2371-100MG
C2371-20MG
TCIAC2371-100MGEA
74
EUR
TCIAC2371-100MG
TCIAC2371-20MG
6-Chloro-3-indolyl-β-D-galactopyranoside ≥98.0% (by HPLC) for biochemical research
6-Chloro-3-indolyl-beta-D-galactopyranoside
Formula:
C₁₄H₁₆ClNO₆ MW: 329.733 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00467206 Núm. CAS: 138182-21-5 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -43.0 to -47.0 deg (C=1, MeOH) |
beta-Galactosidase detection test | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...