1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-(2-Chloroethyl)hexamethyleneimine Hydrochloride, 1-(2-Chloroethyl)azepane hydrochloride
C2379-25G
C2379-5G
TCIAC2379-25GEA
56
EUR
TCIAC2379-25G
TCIAC2379-5G
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride ≥98.0% (by titrimetric analysis)
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride
Formula:
C₈H₁₆ClN MW: 198,13 g/mol |
Número MDL:
MFCD00012842 Núm. CAS: 26487-67-2 EINECS: 247-733-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...