Cyclohexylmethyl acetate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: Acetic Acid Cyclohexylmethyl Ester

C2443-25G C2443-5G
TCIAC2443-25GEA 236 EUR
TCIAC2443-25G TCIAC2443-5G
Cyclohexylmethyl acetate ≥98.0% (por GC)
Cyclohexylmethyl acetate
Formula: C₉H₁₆O₂
MW: 156.23 g/mol
Pto. Ebullición: 183°C
Núm. CAS: 937-55-3

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR