4-Chloro-7-azaindole ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
C2470-200MG
C2470-1G
TCIAC2470-200MGEA
31
EUR
TCIAC2470-200MG
TCIAC2470-1G
4-Chloro-7-azaindole ≥97.0% (by GC, titration analysis)
4-Chloro-7-azaindole
Formula:
C₇H₅ClN₂ MW: 152,58 g/mol |
Número MDL:
MFCD08272232 Núm. CAS: 55052-28-3 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...