Chlorhexidine diacetate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide] diacetate, 1,1'-Hexamethylenebis[5-(4-chlorophenyl)biguanide] Diacetate
C2511-25G
C2511-5G
TCIAC2511-25GEA
132
EUR
TCIAC2511-25G
TCIAC2511-5G
Chlorhexidine diacetate ≥98.0% (by titrimetric analysis)
Chlorhexidine diacetate
Formula:
C₂₂H₃₀Cl₂N₁₀ MW: 625,56 g/mol |
Número MDL:
MFCD00012532 Núm. CAS: 56-95-1 EINECS: 200-302-4 UN: 3077 ADR: 9,III Índice Merck: 14,02091 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min.98.0 % (calcd.on anh.substance) |
Melting point | 153.0 to 160.0 deg-C |
Water | max. 4.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...