4-Carboxy-3-chlorophenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Carboxy-3-chlorobenzeneboronic acid
C2523-1G
C2523-5G
TCIAC2523-1GEA
68
EUR
TCIAC2523-1G
TCIAC2523-5G
4-Carboxy-3-chlorophenylboronic acid (contains varying amounts of Anhydride)
4-Carboxy-3-chlorophenylboronic acid
Formula:
C₇H₆BClO₄ MW: 200,39 g/mol |
Número MDL:
MFCD06656268 Núm. CAS: 136496-72-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...