Clozapine ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]diazepine
C2547-1G
C2547-100MG
TCIAC2547-1GEA
225
EUR
TCIAC2547-1G
TCIAC2547-100MG
Clozapine ≥98.0% (by GC, titration analysis)
Clozapine
Formula:
C₁₈H₁₉ClN₄ MW: 326,83 g/mol |
Número MDL:
MFCD00153785 Núm. CAS: 5786-21-0 EINECS: 227-313-7 UN: 2811 ADR: 6.1,III Índice Merck: 14,02423 |
Resultados test especificaciones
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 183.0 to 187.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...