(-)-Cytisine ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

C2609-1G C2609-100MG
TCIAC2609-1GEA 316 EUR
TCIAC2609-1G TCIAC2609-100MG
(-)-Cytisine ≥98.0% (by GC, titration analysis)
(-)-Cytisine
Formula: C₁₁H₁₄N₂O
Número MDL: MFCD00136048
Núm. CAS: 485-35-8
EINECS: 207-616-0
UN: 2811
ADR: 6.1,III
Índice Merck: 14,02789

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 154.0 to 158.0 deg-C
Specific rotation [a]20/D -109 to -115 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR