N-Carbobenzoxy-S-benzyl-L-cysteine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Cbz-Cys(Bzl)-OH, N-Cbz-S-benzyl-L-cysteine, Z-Cys(Bzl)-OH
C2614-5G
C2614-25G
TCIAC2614-5GEA
29
EUR
TCIAC2614-5G
TCIAC2614-25G
N-Carbobenzoxy-S-benzyl-L-cysteine ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-S-benzyl-L-cysteine
Formula:
C₁₈H₁₉NO₄S MW: 345.41 g/mol |
Número MDL:
MFCD00004785 Núm. CAS: 3257-18-9 EINECS: 221-855-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 96.0 to 100.0 deg-C |
Specific rotation [a]20/D | -44.0 to -48.0 deg (C=4、EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...