1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid
C2650-1G
C2650-5G
TCIAC2650-1GEA
32
EUR
TCIAC2650-1G
TCIAC2650-5G
1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Formula:
C₁₂H₈ClFN₂O₃ MW: 282,6600 g/mol |
Número MDL:
MFCD08458865 Núm. CAS: 100361-18-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 222.0 to 226.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...