Cymoxanil ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Cyan-N-[(ethylamino)carbonyl]-2-(methoxyimino)acetamide, 1-(2-Cyano-2-methoxyiminoacetyl)-3-ethylurea, 2-Cyano-N-(ethylcarbamoyl)-2-methoxyiminoacetamide

C2657-5G C2657-25G
TCIAC2657-5GEA 39 EUR
TCIAC2657-5G TCIAC2657-25G
Cymoxanil ≥98.0% (by HPLC, titration analysis)
Cymoxanil
Formula: C₇H₁₀N₄O₃
MW: 198,18 g/mol
Número MDL: MFCD00137381
Núm. CAS: 57966-95-7
EINECS: 261-043-0
UN: 3077
ADR: 9,III
Índice Merck: 14,02765

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR