3-Chloro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
(3-Chloro-4-ethoxyphenyl)boronic acid
C2664-1G
C2664-5G
TCIAC2664-1GEA
25
EUR
TCIAC2664-1G
TCIAC2664-5G
3-Chloro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-4-ethoxyphenylboronic acid
Formula:
C₈H₁₀BClO₃ MW: 200,4300 g/mol |
Número MDL:
MFCD03701556 Núm. CAS: 279261-81-3 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...