Carprofen ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 2-(6-Chloro-9H-carbazol-2-yl)propanoic acid, 6-Chloro-alpha-methyl-9H-carbazole-2-acetic Acid

C2701-1G C2701-5G
TCIAC2701-1GEA 56 EUR
TCIAC2701-1G TCIAC2701-5G
Carprofen ≥98.0% (by HPLC, titration analysis)
Carprofen
Formula: C₁₅H₁₂ClNO₂
Número MDL: MFCD00079028
Núm. CAS: 53716-49-7
EINECS: 258-712-4
UN: 2811
ADR: 6.1,III
Índice Merck: 14,01862

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 198.0 to 204.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR