2-Chloro-4,6-diphenyl-1,3,5-triazine ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
C2703-5G
C2703-25G
TCIAC2703-5GEA
71
EUR
TCIAC2703-5G
TCIAC2703-25G
2-Chloro-4,6-diphenyl-1,3,5-triazine ≥98.0% (por GC)
2-Chloro-4,6-diphenyl-1,3,5-triazine
Formula:
C₁₅H₁₀ClN₃ MW: 267,72 g/mol |
Número MDL:
MFCD05738885 Núm. CAS: 3842-55-5 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...