1,4-Dibenzyl-L-aspartate-p-toluenesulphonate ≥97.0% (by HPLC, total nitrogen, and titration analysis)
Proveedor: TCI
Sinónimos:
H-Asp(OBzl)-Obzl.TosOH, L-Aspartic Acid Dibenzyl Ester p-Toluenesulphonate Salt, L-Aspartic acid dibenzyl ester p-toluenesulphonate salt Dibenzyl (2S)-2-aminobutanedioate 4-toluenesulphonate, H-Asp(OBzl)-OBzl.Tos-OH, L-Aspartic Acid 1,4-Dibenzyl Ester p-Toluenesulphonate, H-Asp(OBzl)-OBzl·TosOH, L-Aspartic acid dibenzyl ester p-toluenesulphonate
C2711-5G
C2711-25G
TCIAC2711-5GEA
37
EUR
TCIAC2711-5G
TCIAC2711-25G
1,4-Dibenzyl-L-aspartate-p-toluenesulphonate ≥97.0% (by HPLC, total nitrogen, and titration analysis)
1,4-Dibenzyl-L-aspartate-p-toluenesulphonate
Formula:
C₁₈H₁₉NO₄ MW: 485,55 g/mol |
Número MDL:
MFCD00065188 Núm. CAS: 2886-33-1 EINECS: 220-746-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...