Cloruro de 5-cloro-2-metoxibencenosulfonilo ≥95.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
C2740-5G
C2740-25G
TCIAC2740-5GEA
37
EUR
TCIAC2740-5G
TCIAC2740-25G
Cloruro de 5-cloro-2-metoxibencenosulfonilo ≥95.0% (by GC, titration analysis)
Cloruro de 5-cloro-2-metoxibencenosulfonilo
Formula:
C₇H₆Cl₂O₃S MW: 241,09 g/mol |
Número MDL:
MFCD00052959 Núm. CAS: 22952-32-5 UN: 3261 ADR: 8,II |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Argentmetric Titration) | min. 95.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...