7-(2-Chloroethyl)theophylline ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
7-(2-Chloroethyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione
C2748-5G
C2748-25G
TCIAC2748-5GEA
47
EUR
TCIAC2748-5G
TCIAC2748-25G
7-(2-Chloroethyl)theophylline ≥98.0% (by HPLC, titration analysis)
7-(2-Chloroethyl)theophylline
Formula:
C₉H₁₁ClN₄O₂ MW: 242,66 g/mol |
Número MDL:
MFCD00005760 Núm. CAS: 5878-61-5 EINECS: 227-553-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 123.0 to 127.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...