N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
(R)-2-(Carbobenzoxy)amino-2-(4-hydroxyphenyl)acetic Acid, (R)-2-(((Benzyloxy)carbonyl)amino)-2-(4-hydroxyphenyl)acetic acid, Cbz-D-Phg(4-OH)-OH, Z-D-Phg(4-OH)-OH
C2773-25G
C2773-5G
TCIAC2773-25GEA
235
EUR
TCIAC2773-25G
TCIAC2773-5G
N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine
Formula:
C₁₆H₁₅NO₅ MW: 301.30 g/mol |
Número MDL:
MFCD09756650 Núm. CAS: 26787-75-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -124.0 to -129.0 deg (C=10, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...