1-Carbobenzoxy-3-piperidinecarboxylic acid ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-Cbz-3-piperidinecarboxylic Acid, 1-Carbobenzoxynipecotic Acid, 1-Cbz-nipecotic Acid, 1-Carbobenzoxy-3-piperidinecarboxylic acid, 1-[(Benzyloxy)carbonyl]-3-piperidinecarboxylic acid
C2805-1G
C2805-5G
TCIAC2805-1GEA
28
EUR
TCIAC2805-1G
TCIAC2805-5G
1-Carbobenzoxy-3-piperidinecarboxylic acid ≥97.0% (by GC, titration analysis)
1-Carbobenzoxy-3-piperidinecarboxylic acid
Formula:
C₁₄H₁₇NO₄ MW: 263,29 g/mol |
Número MDL:
MFCD02931282 Núm. CAS: 78190-11-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...