4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulphonamide ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-(5-(p-Tolyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl)benzenesulphonamide, Celecoxib
C2816-200MG
C2816-1G
TCIAC2816-200MGEA
21
EUR
TCIAC2816-200MG
TCIAC2816-1G
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulphonamide ≥98.0% (by HPLC, titration analysis)
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulphonamide
Formula:
C₁₇H₁₄F₃N₃O₂S MW: 381,38 g/mol |
Número MDL:
MFCD00941298 Núm. CAS: 169590-42-5 Índice Merck: 14,01956 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 161.0 to 165.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...