4-Chloro-2-(1H-pyrazol-3-yl)phenol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-(5-Chloro-2-hydroxyphenyl)pyrazole
C2818-1G
C2818-5G
TCIAC2818-1GEA
32
EUR
TCIAC2818-1G
TCIAC2818-5G
4-Chloro-2-(1H-pyrazol-3-yl)phenol ≥98.0% (by GC, titration analysis)
4-Chloro-2-(1H-pyrazol-3-yl)phenol
Formula:
C₉H₇ClN₂O MW: 194,62 g/mol |
Número MDL:
MFCD00209566 Núm. CAS: 18704-67-1 EINECS: 242-516-0 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...