Cyclic pifithrin-ɑ-hydrobromide ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
2-(p-Tolyl)-5,6,7,8-tetrahydrobenzo[d]imidazo[2,1-b]thiazole hydrobromide, 2-(p-Tolyl)-5,6,7,8-tetrahydroimidazo[2,1-b]benzothiazole Hydrobromide
C2826-20MG
C2826-100MG
TCIAC2826-20MGEA
241
EUR
TCIAC2826-20MG
TCIAC2826-100MG
Cyclic pifithrin-ɑ-hydrobromide ≥98.0% (by HPLC)
Cyclic pifithrin-alpha-hydrobromide
Formula:
C₁₆H₁₆N₂S Temperatura de almacenaje: Congelador |
Número MDL:
MFCD02683960 Núm. CAS: 511296-88-1 Índice Merck: 14,07421 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...