N-Carbobenzoxy-D-leucine ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: Z-D-Leu-OH, N-Cbz-D-leucine

C2839-5G C2839-25G
TCIAC2839-5GEA 76 EUR
TCIAC2839-5G TCIAC2839-25G
N-Carbobenzoxy-D-leucine ≥97.0% (by HPLC, titration analysis)
N-Carbobenzoxy-D-leucine
Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Número MDL: MFCD00066068
Núm. CAS: 28862-79-5

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D +15.0 to +18.0 deg (C=5, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR