1-(4-Chlorophenyl)-N-methylmethanamine ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
C2885-1G
C2885-5G
TCIAC2885-1GEA
32
EUR
TCIAC2885-1G
TCIAC2885-5G
1-(4-Chlorophenyl)-N-methylmethanamine ≥97.0% (by GC, titration analysis)
1-(4-Chlorophenyl)-N-methylmethanamine
Formula:
C₈H₁₀ClN Pto. Ebullición: 119°C |
Número MDL:
MFCD00018749 Núm. CAS: 104-11-0 EINECS: 203-175-3 UN: 2735 ADR: 8,III |
Resultados test especificaciones
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...