5-Chlorobenzofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

C2890-1G C2890-5G
TCIAC2890-1GEA 53 EUR
TCIAC2890-1G TCIAC2890-5G
5-Chlorobenzofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)
5-Chlorobenzofuran-2-carboxylic acid
Formula: C₉H₅ClO₃
Número MDL: MFCD00060512
Núm. CAS: 10242-10-1

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR