1-Cyclopentenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
Cyclopent-1-en-1-ylboronic acid
C2893-1G
C2893-5G
TCIAC2893-1GEA
37
EUR
TCIAC2893-1G
TCIAC2893-5G
1-Cyclopentenylboronic acid (contains varying amounts of Anhydride)
1-Cyclopentenylboronic acid
Formula:
C₅H₉BO₂ MW: 111,94 g/mol |
Número MDL:
MFCD02179496 Núm. CAS: 850036-28-1 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | 93.0 to 120.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...