1-(3-Chloropropyl)piperidine hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: N-(3-Chloropropyl)piperidine hydrochloride

C2922-1G C2922-5G
TCIAC2922-1GEA 28 EUR
TCIAC2922-1G TCIAC2922-5G
1-(3-Chloropropyl)piperidine hydrochloride ≥98.0% (by titrimetric analysis)
1-(3-Chloropropyl)piperidine hydrochloride
Formula: C₈H₁₇Cl₂N
MW: 198,14 g/mol
Número MDL: MFCD00012838
Núm. CAS: 5472-49-1
EINECS: 226-812-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 216.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR