3-Cyano-4-methoxy-2-pyridone ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
3-Cyano-2-hydroxy-4-methoxypyridine, 2-Hydroxy-4-methoxynicotinonitrile, 3-Cyano-4-methoxy-2-pyridinone
C2925-5G
C2925-25G
TCIAC2925-5GEA
53
EUR
TCIAC2925-5G
TCIAC2925-25G
3-Cyano-4-methoxy-2-pyridone ≥98.0% (by HPLC, titration analysis)
3-Cyano-4-methoxy-2-pyridone
Formula:
C₇H₆N₂O₂ MW: 150,14 g/mol |
Número MDL:
MFCD08059559 Núm. CAS: 21642-98-8 UN: 3439 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...