Chlorambucil ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
4-(4-(Bis(2-chloroethyl)amino)phenyl)butanoic acid, 4-[4-[Bis(2-chloroethyl)amino]phenyl]butyric Acid
C2939-200MG
C2939-1G
TCIAC2939-200MGEA
50
EUR
TCIAC2939-200MG
TCIAC2939-1G
Chlorambucil ≥98.0% (by GC, titration analysis)
Chlorambucil
Formula:
C₁₄H₁₉Cl₂NO₂ MW: 304,21 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00021783 Núm. CAS: 305-03-3 EINECS: 206-162-0 UN: 2811 ADR: 6.1,III Índice Merck: 14,02073 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...