Cyproterone acetate ≥98.0% (by HPLC)
Proveedor: TCI
|
Peligro
|
C2942-200MG
C2942-1G
TCIAC2942-200MGEA
70
EUR
TCIAC2942-200MG
TCIAC2942-1G
Cyproterone acetate ≥98.0% (by HPLC)
Cyproterone acetate
Formula:
C₂₄H₂₉ClO₄ MW: 416.94 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00864671 Núm. CAS: 427-51-0 EINECS: 207-048-3 Índice Merck: 14,02774 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 96.0 % |
Melting point | 206.0 to 211.0 deg-C |
Specific rotation [a]20/D | +148.0 to +156.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...