3-Carboxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 3-Carboxy-4-fluorophenylboronic Acid, 3-Carboxy-4-fluorobenzeneboronic acid

C2945-1G C2945-5G
TCIAC2945-1GEA 46 EUR
TCIAC2945-1G TCIAC2945-5G
3-Carboxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Carboxy-4-fluorophenylboronic acid
Formula: C₇H₆BFO₄
MW: 183,93 g/mol
Número MDL: MFCD03095129
Núm. CAS: 872460-12-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR