4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-iodoquinazolin-4-amine

C2965-1G C2965-5G
TCIAC2965-1GEA 40 EUR
TCIAC2965-1G TCIAC2965-5G
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline ≥98.0% (by HPLC, titration analysis)
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline
Formula: C₂₁H₁₄ClFIN₃O
MW: 505,72 g/mol
Número MDL: MFCD09998827
Núm. CAS: 231278-20-9

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 223.0 to 227.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR