N-(3-Chlorophenyl)-3-oxobutyramide ≥96.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

C2984-200MG C2984-1G
TCIAC2984-200MGEA 55 EUR
TCIAC2984-200MG TCIAC2984-1G
N-(3-Chlorophenyl)-3-oxobutyramide ≥96.0% (by HPLC, titration analysis)
N-(3-Chlorophenyl)-3-oxobutyramide
Formula: C₁₀H₁₀ClNO₂
Número MDL: MFCD00018440
Núm. CAS: 2415-87-4

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 96.0 %
Melting point 101.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR