Cephradine monohydrate ≥96.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

C2988-1G C2988-5G
TCIAC2988-1GEA 59 EUR
TCIAC2988-1G TCIAC2988-5G
Cephradine monohydrate ≥96.0% (by HPLC, titration analysis)
Cephradine monohydrate
Formula: C₁₆H₁₉N₃O₄S·H₂O
Temperatura de almacenaje: Nevera
Número MDL: MFCD00865048
Núm. CAS: 75975-70-1
EINECS: 254-137-8
Índice Merck: 14,01986

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +85.0 to +89.0 deg (C=0.5, H2O) (calcd.on anh.substance)
Water 3.0 to 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR