(S)-β-(Carbobenzoxyamino)-γ-butyrolactone ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: (S)-Benzyl (5-oxotetrahydrofuran-3-yl)carbamate, Benzyl (S)-Tetrahydro-5-oxo-3-furanylcarbamate, (S)-4-(Carbobenzoxyamino)tetrahydrofuran-2-one, (S)-beta-(Cbz-amino)-gamma-butyrolactone, (S)-4-(Cbz-amino)tetrahydrofuran-2-one

C2995-1G C2995-5G
TCIAC2995-1GEA 23 EUR
TCIAC2995-1G TCIAC2995-5G
(S)-β-(Carbobenzoxyamino)-γ-butyrolactone ≥98.0% (by HPLC, total nitrogen)
(S)-beta-(Carbobenzoxyamino)-gamma-butyrolactone
Formula: C₁₂H₁₃NO₄
MW: 235,24 g/mol
Número MDL: MFCD00216569
Núm. CAS: 87219-29-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 103.0 to 107.0 deg-C
Specific rotation [a]20/D -53.0 to -57.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR