Combretastatin A4 Phosphate disodium salt ≥98.0% (by HPLC)
Proveedor: TCI
C3009-20MG
C3009-100MG
TCIAC3009-20MGEA
93
EUR
TCIAC3009-20MG
TCIAC3009-100MG
Combretastatin A4 Phosphate disodium salt ≥98.0% (by HPLC)
Combretastatin A4 Phosphate disodium salt
Formula:
C₁₈H₁₉Na₂O₈P Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00943911 Núm. CAS: 168555-66-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...