Chlormethiazole hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 5-(2-Chloroethyl)-4-methylthiazole Hydrochloride, Somnevrin Hydrochloride, Clomethiazole Hydrochloride

C3020-200MG C3020-1G
TCIAC3020-200MGEA 74 EUR
TCIAC3020-200MG TCIAC3020-1G
Chlormethiazole hydrochloride ≥98.0% (by HPLC, titration analysis)
Chlormethiazole hydrochloride
Formula: C₆H₈ClNS
MW: 198,11 g/mol
Número MDL: MFCD00673948
Núm. CAS: 6001-74-7
Índice Merck: 14,02384

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 134.0 to 138.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR