Carbonylhydridotris(triphenylphosphine)iridium(I)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Hydridocarbonyltris(triphenylphosphine)iridium(I)
C3040-200MG
C3040-1G
TCIAC3040-200MGEA
171
EUR
TCIAC3040-200MG
TCIAC3040-1G
Carbonylhydridotris(triphenylphosphine)iridium(I)
Carbonylhydridotris(triphenylphosphine)iridium(I)
Formula:
C₅₅H₄₆IrOP₃ MW: 1008,12 g/mol |
Número MDL:
MFCD00015525 Núm. CAS: 17250-25-8 EINECS: 241-282-7 UN: 2811 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Yellow to Green powder to crystal |
Elemental analysis (Carbon) | 62.00 to 69.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...