Coumarin 504T ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: Ethyl 2,3,6,7-Tetrahydro-1,1,7,7-tetramethyl-11-oxo-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizine-10-carboxylate, Ethyl 1,1,7,7-tetramethyl-11-oxo-2,3,5,6,7,11-hexahydro-1H-pyrano[2,3-f]pyrido[3,2,1-ij]quinoline-10-carboxylate

C3045-200MG C3045-1G
TCIAC3045-200MGEA 68 EUR
TCIAC3045-200MG TCIAC3045-1G
Coumarin 504T ≥98.0% (by HPLC, total nitrogen)
Coumarin 504T
Formula: C₂₂H₂₇NO₄
MW: 369,46 g/mol
Número MDL: MFCD00144013
Núm. CAS: 113869-06-0

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 127.0 to 131.0 deg-C
Lambda max. 430.0 to 436.0 nm (EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR