Ácido 3-cloro-4-metoxifenilborónico (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 3-Chloro-4-methoxybenzeneboronic acid, (3-Chloro-4-methoxyphenyl)boronic acid

C3056-1G C3056-5G
TCIAC3056-1GEA 39 EUR
TCIAC3056-1G TCIAC3056-5G
Ácido 3-cloro-4-metoxifenilborónico (contains varying amounts of Anhydride)
Ácido 3-cloro-4-metoxifenilborónico
Formula: C₇H₈BClO₃
MW: 186,40 g/mol
Número MDL: MFCD04039888
Núm. CAS: 175883-60-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR