2-Cloro-6-nitrofenol ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

C3074-1G C3074-5G
TCIAC3074-1GEA 27 EUR
TCIAC3074-1G TCIAC3074-5G
2-Cloro-6-nitrofenol ≥98.0% (by GC, titration analysis)
2-Cloro-6-nitrofenol
Formula: C₆H₄ClNO₃
Número MDL: MFCD00024233
Núm. CAS: 603-86-1
EINECS: 210-061-7

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR