4-Clorofenilamidina clorhidrato ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 4-Chlorophenylamidine hydrochloride, 4-Chlorobenzene-1-carboximidamide hydrochloride

C3090-5G C3090-25G
TCIAC3090-5GEA 35 EUR
TCIAC3090-5G TCIAC3090-25G
4-Clorofenilamidina clorhidrato ≥98.0% (by HPLC, titration analysis)
4-Clorofenilamidina clorhidrato
Formula: C₇H₈Cl₂N₂
MW: 191,06 g/mol
Número MDL: MFCD00126401
Núm. CAS: 14401-51-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 244.0 to 246.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR