3-Chloro-4-methyl-7-hydroxycoumarin ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 3-Chloro-4-methylumbelliferone, 3-Chloro-7-hydroxy-4-methylcoumarin

C3098-1G C3098-5G
TCIAC3098-1GEA 71 EUR
TCIAC3098-1G TCIAC3098-5G
3-Chloro-4-methyl-7-hydroxycoumarin ≥98.0% (by titrimetric analysis)
3-Chloro-4-methyl-7-hydroxycoumarin
Formula: C₁₀H₇ClO₃
MW: 210,61 g/mol
Número MDL: MFCD00006851
Núm. CAS: 6174-86-3
EINECS: 228-217-8

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 241.0 to 245.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR