4-Cloro-3-fluorobenzonitrilo ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
C3103-5G
C3103-1G
TCIAC3103-5GEA
124
EUR
TCIAC3103-5G
TCIAC3103-1G
4-Cloro-3-fluorobenzonitrilo ≥98.0% (por GC)
4-Cloro-3-fluorobenzonitrilo
Formula:
C₇H₃ClFN |
Número MDL:
MFCD00143291 Núm. CAS: 110888-15-8 UN: 3439 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...