2-Chloro-5-fluoro-3-nitropyridine ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
C3115-200MG
C3115-1G
TCIAC3115-200MGEA
48
EUR
TCIAC3115-200MG
TCIAC3115-1G
2-Chloro-5-fluoro-3-nitropyridine ≥98.0% (por GC)
2-Chloro-5-fluoro-3-nitropyridine
Formula:
C₅H₂ClFN₂O₂ MW: 176,5300 g/mol |
Número MDL:
MFCD06659490 Núm. CAS: 136888-21-6 |
Resultados test especificaciones
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...