4-Chloro-2,5-xylidine ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Chloro-2,5-dimethylaniline
C3145-1G
C3145-5G
TCIAC3145-1GEA
32
EUR
TCIAC3145-1G
TCIAC3145-5G
4-Chloro-2,5-xylidine ≥98.0% (by GC, titration analysis)
4-Chloro-2,5-xylidine
Formula:
C₈H₁₀ClN MW: 155,63 g/mol |
Número MDL:
MFCD08445705 Núm. CAS: 20782-94-9 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 91.0 to 95.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...