2-(3'-Chlorobiphenyl-4-yl)-4,6-diphenyl-1,3,5-triazine ≥97.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(3'-Chloro-[1,1'-biphenyl]-4-yl)-4,6-diphenyl-1,3,5-triazine
C3163-200MG
C3163-1G
TCIAC3163-200MGEA
39
EUR
TCIAC3163-200MG
TCIAC3163-1G
2-(3'-Chlorobiphenyl-4-yl)-4,6-diphenyl-1,3,5-triazine ≥97.0% (by HPLC, total nitrogen)
2-(3'-Chlorobiphenyl-4-yl)-4,6-diphenyl-1,3,5-triazine
Formula:
C₂₇H₁₈ClN₃ MW: 419,91 g/mol |
Número MDL:
MFCD28975107 Núm. CAS: 1443049-84-0 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 204.0 to 208.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...