1-(3-Clorofenil)-2-tiourea ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Peligro
|
Sinónimos:
3-Clorofeniltiourea, N-(3-Clorofenil)tiourea
C3168-1G
C3168-5G
TCIAC3168-1GEA
23
EUR
TCIAC3168-1G
TCIAC3168-5G
1-(3-Clorofenil)-2-tiourea ≥98.0% (by HPLC, total nitrogen)
1-(3-Clorofenil)-2-tiourea
Formula:
(C₆H₄Cl)NC(S)NH₂ MW: 186.66 g/mol Pto. Ebullición: 304 |
Número MDL:
MFCD00022165 Núm. CAS: 4947-89-1 UN: 2811 ADR: 6.1 II |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 137.0 to 141.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...