2-Chloro-5-methylphenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
(2-Chloro-5-methylphenyl)boronic acid, 2-Chloro-5-methylbenzeneboronic acid
C3211-1G
C3211-5G
TCIAC3211-1GEA
80
EUR
TCIAC3211-1G
TCIAC3211-5G
2-Chloro-5-methylphenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-5-methylphenylboronic acid
Formula:
C₇H₈BClO₂ MW: 170,40 g/mol |
Número MDL:
MFCD06659822 Núm. CAS: 193353-35-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...